Tetrabutylammoniumtetraphenylborat, Supelco®

Lieferant: Merck

Synonyme: Tetrabutylammonium Tetraphenylborate

M-Clarity™ Qualitätsstufe = MQ100

86897-5G
86897-5GEA 495 EUR
86897-5G
Tetrabutylammoniumtetraphenylborat, Supelco®
Tetrabutylammonium tetraphenylborate
Formel: C₄₀H₅₆BN
Molecular Weight: 561,6904 g/mol
MDL: MFCD00011749
CAS-Nummer: 15522-59-5
EINECS: 239-562-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff6a0b04fcbea8a5f37ae9aaffe390e344c24b6&vid=7f06302bda45ecbe0182d88b695265f14355dfa5&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR