Cobalt(II)nitrat Hexahydrat ≥99,0% ACS, Reag. Ph. Eur. (max.0.001% Ni)
Lieferant: VWR Chemicals
|
Gefahr
|
Synonyme:
Cobaltnitrat Hexahydrat, Cobaltdinitrat Hexahydrat
87027.230EA
526
EUR
87027.230
Cobalt(II)nitrat Hexahydrat ≥99,0% ACS, Reag. Ph. Eur. (max.0.001% Ni)
Cobalt(II)nitrat Hexahydrat
Formel:
Co(NO₃)₂·6H₂O Molecular Weight: 291,03 g/mol Schmelzpunkt: 57 °C Dichte: 1,88 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149647 CAS-Nummer: 10026-22-9 EINECS: 233-402-1 UN: 1477 ADR: 5.1,III Merck Index: 13,02469 |
Spezifikation Testergebnisse
Assay (Complexometric) | 99,0 - 102,0 |
Identity (IR-Spectrum) | passes test |
Insoluble in water | ≤ 0,005 % |
Chlorides (Cl) | ≤ 0,002 % |
Sulfates (SO4) | ≤ 0,005 % |
Ammonium (NH4) | ≤ 0,05 % |
Calcium (Ca) | ≤ 0,005 % |
Copper (Cu) | ≤ 0,001 % |
Iron (Fe) | ≤ 0,001 % |
Lead (Pb) | ≤ 0,001 % |
Magnesium (Mg) | ≤ 0,005 % |
Manganese (Mn) | ≤ 0,005 % |
Nickel (Ni) | ≤ 0,001 % |
Potassium (K) | ≤ 0,01 % |
Sodium (Na) | ≤ 0,05 % |
Zinc (Zn) | ≤ 0,005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...