Benzoylchlorid ≥98,0%, GPR RECTAPUR®

Lieferant: VWR Chemicals

87035.290EA 67.6 EUR
87035.290 87035.460
Benzoylchlorid ≥98,0%, GPR RECTAPUR®
Benzoylchlorid
Formel: C₇H₅ClO
Lagertemperatur: Raumtemperatur
MDL: MFCD00000653
CAS-Nummer: 98-88-4
UN: 1736
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear liquid
Assay
Colour (APHA) ≤50
Identity Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR