Cyclohexan, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Gefahr

8706.1000EA 0 EUR
8706.1000 8706.2500
Cyclohexan, BAKER ANALYZED® ACS, J.T.Baker®
Cyclohexan
Formel: C₆H₁₂
Siedepunkt: 81 °C (1013 hPa)
Schmelzpunkt: 7 °C
Dichte: 0,7785 g/cm³ (20 °C)
Flammpunkt: –18 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003814
CAS-Nummer: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets ACS Specifications
Assay (by GC) min. 99.0 %
Color (APHA) max. 10
Residue after Evaporation max. 0.002 %
Substances Darkened by H2SO4 passes test
Water (H2O) max. 0.02 %
Ultraviolet Absorbance (1.00-cm path vs water):
at 210 nm max. 1.00
at 220 nm max. 0.50
at 230 nm max. 0.20
at 240 nm max. 0.08
at 250 nm max. 0.03
at 260 nm max. 0.02
at 300-400 nm max. 0.01


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR