2,6-Di-tert-butyl-p-kresol ≥99%, GPR RECTAPUR®
Lieferant: VWR Chemicals
|
Achtung
|
87069.460EA
0
EUR
87069.460
2,6-Di-tert-butyl-p-kresol ≥99%, GPR RECTAPUR®
2,6-Di-tert-butyl-p-kresol
Formel:
C₁₅H₂₄O |
MDL:
MFCD00011644 CAS-Nummer: 128-37-0 UN: 3077 ADR: 9,III |
Spezifikation Testergebnisse
Assay | Min. 99 % |
Identification (IR) | Conforms to structure |
Freezing point | 69.2 - 70.0 °C |
Heavy metals (as Pb) | Max. 10 ppm |
Ignition residue | Max. 20 ppm |
Ignition residue (SO4) | Max. 0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...