HMDS (1,1,1,3,3,3-Hexamethyldisilazan) ≥98%, GPR RECTAPUR®

Lieferant: VWR Chemicals

Synonyme: Bis(trimethylsilyl)amin

87078.150EA 24.8 EUR
87078.150
HMDS (1,1,1,3,3,3-Hexamethyldisilazan) ≥98%, GPR RECTAPUR®
HMDS (1,1,1,3,3,3-Hexamethyldisilazan)
Formel: (CH₃)₃SiNHSi(CH₃)₃
Molecular Weight: 161,39 g/mol
MDL: MFCD00008259
CAS-Nummer: 999-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce35a38a4179292c4ff37b2f3093018738545e6e&vid=1693ad54a45ca3ae50a06056893e8438d1002549&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay 98
Density (20/4) 0.774 - 0.775

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR