n-Tetradecan ≥99,0%, GPR RECTAPUR®

Lieferant: VWR Chemicals
Gefahr

Synonyme: Tetradecane

87079.230EA 135 EUR
87079.230
n-Tetradecan ≥99,0%, GPR RECTAPUR®
n-Tetradecan
Formel: H₃C(CH₂)₁₂CH₃
Molecular Weight: 198,39 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008986
CAS-Nummer: 629-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03064a29015d1e810f1e4350a7a6cd2dfe4713fd&vid=f4838a5686fe033622495a3bf280963555654d9a&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay Min. 99,0%
Identification (IR) Passes test
Density (25/4) 0,761 - 0,765

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR