Bortrifluorid-Diethylether-Komplex, GPR RECTAPUR® zur Synthese

Lieferant: VWR Chemicals
Gefahr

87083.230EA 44.5 EUR
87083.230
Bortrifluorid-Diethylether-Komplex, GPR RECTAPUR® zur Synthese
Bortrifluorid-Diethylether-Komplex
Formel: C₄H₁₀BF₃O
MDL: MFCD00013194
CAS-Nummer: 109-63-7
EINECS: 203-689-8
UN: 2604
ADR: 8 (3),I

Bestellen

Spezifikation Testergebnisse

Assay (calculated as BF₃) 45 - 50 %
Density (20/4) 1,124 - 1,127
Refractive index (20 °C; 589 nm) 1,343 - 1,344


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR