Tetraethylammoniumbromid ≥99%, GPR RECTAPUR®
Lieferant: VWR Chemicals
|
Achtung
|
Synonyme:
Tetrylammoniumbromid
87084.290EA
113
EUR
87084.290
Tetraethylammoniumbromid ≥99%, GPR RECTAPUR®
Tetraethylammoniumbromid
Formel:
C₈H₂₀BrN Molecular Weight: 210,16 g/mol Schmelzpunkt: 285 °C |
MDL:
MFCD00011825 CAS-Nummer: 71-91-0 |
Spezifikation Testergebnisse
Assay | Min. 99% |
Water | Max. 1,0% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...