Benzylchlorid ≥99%, GPR RECTAPUR®

Lieferant: VWR Chemicals

Synonyme: Chlormethylbenzol, α-Chlortoluol, Chlorphenylmethan, (Chlormethyl)benzol

87103.320EA 98.3 EUR
87103.320
Benzylchlorid ≥99%, GPR RECTAPUR®
Benzylchlorid
Formel: (C₆H₅)CH₂Cl
Lagertemperatur: Raumtemperatur
MDL: MFCD00000889
CAS-Nummer: 100-44-7
UN: 1738
ADR: 6.1 (8),II

Bestellen

Spezifikation Testergebnisse

Assay (GC, area%) ≥ 99%
Appearance: Clear colourless to pale yellow liquid Passes test
Density (20 °C) 1,097 - 1,101
Acidity ≤ 0,002 meq/g
Non-volatile matter ≤ 0,05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR