Anisol ≥99%, GPR RECTAPUR®
Lieferant: VWR Chemicals
|
Gefahr
|
87106.260EA
41.8
EUR
87106.260
87106.320
87106.460
Anisol ≥99%, GPR RECTAPUR®
Anisol
Formel:
C₇H₈O Flammpunkt: 52 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008354 CAS-Nummer: 100-66-3 UN: 2222 ADR: 3,III |
Spezifikation Testergebnisse
Assay | ≥ 99 % |
Appearance | Clear colourless liquid |
Odour | Characteristic |
Density (20/4) | 0,992 - 0,994 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...