1-Tetradecen, Supelco®

Lieferant: Merck

87187-5ML
87187-5MLEA 116 EUR
87187-5ML
1-Tetradecen, Supelco®
1-Tetradecen
Formel: C₁₄H₂₈
MDL: MFCD00008981
CAS-Nummer: 1120-36-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=200a2e4c0244030e459c4c087bfc6a0ef7d54fea&vid=d21261b477d77359c818aeae3e442e36f5951e61&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR