Copper sulphate-Lösung

Lieferant: VWR Chemicals

Synonyme: Copper(II) sulphate, Cupric sulphate

87204.360EA 189 EUR
87204.360
Copper sulphate-Lösung
Copper(II) sulphate solutions
Formel: CuSO₄·5H₂O
Molecular Weight: 249,69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00149681
CAS-Nummer: 7758-99-8

Bestellen

Spezifikation Testergebnisse

Density 1.165 - 1.195

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR