Tetradecyltrimethylammoniumbromid, Supelco®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Myristyltrimethylammoniumbromid, Tetradoniumbromid, TTAB, Dynamin Inhibitor II
M-Clarity™ Qualitätsstufe = MQ100
87208-10G
87208-10GEA
275
EUR
87208-10G
Tetradecyltrimethylammoniumbromid, Supelco®
Tetradecyltrimethylammoniumbromid
Formel:
C₁₇H₃₈BrN Molecular Weight: 336,394 g/mol Schmelzpunkt: 245…250 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011770 CAS-Nummer: 1119-97-7 EINECS: 214-291-9 UN: 3261 ADR: 8,II Merck Index: 12,06419 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...