Tetradodecylammonium nitrate ≥99,0%, Selectophore™ Ionophor, Supelco®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

87252-100MG
87252-100MGEA 63.8 EUR
87252-100MG
Tetradodecylammonium nitrate ≥99,0%, Selectophore™ Ionophor, Supelco®
Tetradodecylammonium nitrate

Tetradodecylammonium nitrate (TDDAN) is a quaternary ammonium salt, mainly used as electrolyte.


TDDAN has been used as solvent in the preparation of microelectrodes based on AgI-Ag2O-V2O5 glass transducer.


Ion-pair suitable for a nitrate-selective field-effect transistor membrane.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: [CH₃(CH₂)₁₀CH₂]₄N(NO₃)
Molecular Weight: 753,32 g/mol
MDL: MFCD00467971
CAS-Nummer: 63893-35-6
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ed4fcf725b6fede982e1fd542c0ef62a077f261&vid=d253e3486b12b89f925084c0fe165864d0c10548&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR