Tetradodecylammonium tetrakis(4-chlorophenyl)borate, Selectophore™ Ionophor, Supelco®

Lieferant: Merck
Achtung

Synonyme: ETH 500, Tetrakis(4-chlorophenyl)borate tetradodecylammonium salt

M-Clarity™ Qualitätsstufe = MQ100

87255-1G
87255-1GEA 258 EUR
87255-1G
Tetradodecylammonium tetrakis(4-chlorophenyl)borate, Selectophore™ Ionophor, Supelco®
Tetradodecylammonium tetrakis(4-chlorophenyl)borate

ETH 500 is a lipophilic electrolyte mostly used as an anionic additive.


ETH 500 has been used in the preparation of molecularly imprinted polymers, which can be used during potentiometric sensing method for determining neutral phenols.

Formel: C₇₂H₁₁₆BCl₄N
Molecular Weight: 1148.32 g/mol
MDL: MFCD00145372
CAS-Nummer: 100581-42-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=359120219ddc41cb19cca370dcf19f3dd9760e38&vid=77377f0f10ff4a8b129f5e3a3b8200a6e5473ca8&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR