Tetraethylammonium benzoate ≥99,0% zur elektrochemischen Analyse, Supelco®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

87259-5G
87259-5GEA 140 EUR
87259-5G
Tetraethylammonium benzoate ≥99,0% zur elektrochemischen Analyse, Supelco®
Tetraethylammonium benzoate
Formel: (CH₃CH₂)₄N(C₆H₅COO)
MDL: MFCD00041976
CAS-Nummer: 16909-22-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f02184f2243d2b548ccc12e73e2733537a294&vid=e3bb53814c1217f5ba4eaa0b600784e291c5cf8e&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR