Tetrahexylammoniumbromid, Supelco®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
87297-10G-F
87297-10G-FEA
318
EUR
87297-10G-F
Tetrahexylammoniumbromid, Supelco®
Tetrahexylammoniumbromid
Formel:
C₂₄H₅₂BrN Molecular Weight: 434,59 g/mol Schmelzpunkt: 104 °C Dichte: 1,0036 g/cm³ (25 °C) |
MDL:
MFCD00011858 CAS-Nummer: 4328-13-6 EINECS: 224-363-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...