Tetrahydroxyquinone dihydrate Indikator für Titrationen (Ba, SO4), Fluka™

Lieferant: Honeywell Chemicals
Achtung

Synonyme: Tetrahydroxy-1,4-benzenequinone dihydrate

87510-5G
87510-5GEA 365.96 EUR
87510-5G
Tetrahydroxyquinone dihydrate Indikator für Titrationen (Ba, SO4), Fluka™
Tetrahydroxyquinone dihydrate
Formel: C₆H₄O₆·₂H₂O
Molecular Weight: 208.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00001597
CAS-Nummer: 5676-48-2

Bestellen

Spezifikation Testergebnisse

Ignition Residue ≤0.5%
Identity (IR) complying
Assay (calculated to the dried substance) 98.5 - 101.5 %
Colour of the solution complying
Clearness of the solution complying
Residue on ignition Max. 0.5 %
Application test complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR