Magnesiumnitrat Hexahydrat ≥99,97% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99.97%
CAS No.: 13446-18-9
Molecular Formula: Mg(NO3)2.6H2O
Formula Weight: 256.41 (148.32anhy)
Physical Form: Crystalline
Density: 1.46
Melting Point: 89°
Boiling Point: 330° dec.
Merck Reference: 14,5674
UN No.: 1474
MDL No.: MFCD00149779
Formel:
Mg(NO₃)₂·6H₂O Siedepunkt: 330 °C (Zersetzung) Schmelzpunkt: 89 °C Dichte: 1,46 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149779 CAS-Nummer: 13446-18-9 EINECS: 233-826-7 UN: 1474 ADR: 5.1,III Merck Index: 14,5674 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...