Natriumborhydrid 4.4 M in wässriger Lösung stabil
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
87659.AE
87659.AP
87659.K3
87659.AEEA
31.7
EUR
87659.AE
87659.AP
87659.K3
Natriumborhydrid 4.4 M in wässriger Lösung stabil
Natriumtetrahydroborat
CAS No.: 16940-66-2
Molecular Formula: BH4Na
Formula Weight: 37.83
Physical Form: Liquid
Density: 1.4
Sensitivity: Air Sensitive
Appearance: Colourless
UN No.: UN3320
MDL No.: MFCD00003518
Formel:
NaBH₄ Dichte: 1,4 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003518 CAS-Nummer: 16940-66-2 EINECS: 241-004-4 UN: 3320 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...