Methylrot-Lösung Reag. Ph. Eur. 1055101

Lieferant: VWR Chemicals

Synonyme: 2-(4-(Dimethylamino)phenylazo)benzoesäure

87703.180EA 52.5 EUR
87703.180
Methylrot-Lösung Reag. Ph. Eur. 1055101
Methylrot
Formel: C₁₅H₁₅N₃O₂
Molecular Weight: 269,3 g/mol
MDL: MFCD00002425
CAS-Nummer: 493-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR