Phenolphthalein 0,1% in Ethanol 80% Reag. Ph. Eur. 1063702

Lieferant: VWR Chemicals

Synonyme: 3,3-Bis(4-hydroxyphenyl)phthalid, 3,3-Bis(4-hydroxyphenyl)-1(3H)-isobenzofuranon

87705.180EA 56.8 EUR
87705.180
Phenolphthalein 0,1% in Ethanol 80% Reag. Ph. Eur. 1063702
Phenolphthalein

Bestellinformation: Normally manufactured to order. Please check with customer services.

Formel: C₂₀H₁₄O₄
Molecular Weight: 318,33 g/mol
MDL: MFCD00005913
CAS-Nummer: 77-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR