Cobalt(II)oxalat Dihydrat, Reagenzqualität
Lieferant: Thermo Fisher Scientific
|
Achtung
|
CAS No.: 5965-38-8
Molecular Formula: CoC2O4.2H2O
Formula Weight: 182.98 (146.95anhy)
Physical Form: Powder
Density: 3.021
Odour: Odourless
Merck Reference: 14,2445
UN No.: 3288
MDL No.: MFCD00050793
Stabilizer for HCN, temperature indicator
Formel:
C₂CoO₄·2H₂O Schmelzpunkt: 229…231 °C (Zersetzung) Dichte: 3,021 Lagertemperatur: Raumtemperatur |
MDL:
MFCD03414148 CAS-Nummer: 5965-38-8 EINECS: 212-409-3 UN: 3288 ADR: 6.1,III Merck Index: 14,2445 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...