Bromkresolgrün Natriumsalz 0,5 g/l in Ethanol 20% (v/v) Reag. Ph. Eur. 1012601

Lieferant: VWR Chemicals

Synonyme: 3,3',5,5'-Tetrabrom-m-kresolsulfonphthalein Natriumsalz, Bromkresolgrün wasserlöslich

87791.180EA 52.5 EUR
87791.180
Bromkresolgrün Natriumsalz 0,5 g/l in Ethanol 20% (v/v) Reag. Ph. Eur. 1012601
Bromkresolgrün Natriumsalz

Bestellinformation: Normally manufactured to order. Please check with customer services.

Formel: C₂₁H₁₃Br₄NaO₅S
Molecular Weight: 720 g/mol
MDL: MFCD00148898
CAS-Nummer: 62625-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR