Bromkresolpurpur -Lösung Reag. Ph. Eur. 1012701
Lieferant: VWR Chemicals
Synonyme:
5′,5''-Dibrom-o-kresolsulfonphthalein
87793.180EA
52.5
EUR
87793.180
Bromkresolpurpur -Lösung Reag. Ph. Eur. 1012701
Bromkresolpurpur
Formel:
C₂₁H₁₆Br₂O₅S Molecular Weight: 540.23 g/mol Schmelzpunkt: 242 |
MDL:
MFCD00011681 CAS-Nummer: 115-40-2 Merck Index: 13,01371 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...