Bromphenolblau -Lösung Reag. Ph. Eur. 1012801

Lieferant: VWR Chemicals

Synonyme: 3,3',5,5'-Tetrabromphenolsulfonphthalein, 4,4'-(3H-2,1-Benzoxathiol-3-yliden)bis[2,6-dibromphenol]-S,S-dioxid

87794.180EA 52.5 EUR
87794.180
Bromphenolblau -Lösung Reag. Ph. Eur. 1012801
Bromphenolblau
Formel: C₁₉H₁₀Br₄O₅S
MDL: MFCD00005875
CAS-Nummer: 115-39-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR