Kongorot 0,1% in Ethanol 15% (v/v) Reag. Ph. Eur. 1022001

Lieferant: VWR Chemicals

Synonyme: Direktrot 28, Direct Red 28, Dinatrium-3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalin-1-sulfonat)

87805.180EA 52.5 EUR
87805.180
Kongorot 0,1% in Ethanol 15% (v/v) Reag. Ph. Eur. 1022001
Kongorot

Bestellinformation: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: C₃₂H₂₂N₆Na₂O₆S₂
Molecular Weight: 696,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004028
CAS-Nummer: 573-58-0
EINECS: 209-358-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR