Kresolrot Reag. Ph. Eur. 1022801
Lieferant: VWR Chemicals
Synonyme:
o-Cresolsulphonphthalein, 3,3-Bis(4-hydroxy-3-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
87810.180EA
52.5
EUR
87810.180
Kresolrot Reag. Ph. Eur. 1022801
Kresolrot
Formel:
C₂₁H₁₈O₅S Molecular Weight: 382,43 g/mol |
MDL:
MFCD00005878 CAS-Nummer: 1733-12-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...