Kaliumhydroxid 6,6 g/l in Ethanol 95% Reag. Ph. Eur. 1070304

Lieferant: VWR Chemicals

Synonyme: Kalihydrat, Ätzkali

87907.290EA 111 EUR
87907.290
Kaliumhydroxid 6,6 g/l in Ethanol 95% Reag. Ph. Eur. 1070304
Kaliumhydroxid

Bestellinformation: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: KOH
Molecular Weight: 56.11 g/mol
Siedepunkt: 1320
Schmelzpunkt: 360
MDL: MFCD00003553
CAS-Nummer: 1310-58-3
ADR: 3 II
Merck Index: 12,07806

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR