Thymolblau Natriumsalz 0,1% in Ethanol 20% Reag. Ph. Eur. 1090601
Lieferant: VWR Chemicals
Synonyme:
Thymolsulfonphthalein Natriumsalz, Natriumhydrogen-4,4'-(3H-2,1-benzoxathiol-3-yliden)bis[5-methyl-2-(1-methylethyl)phenolat]-S,S-dioxid, Thymolblau wasserlöslich
87952.180EA
52.5
EUR
87952.180
Thymolblau Natriumsalz 0,1% in Ethanol 20% Reag. Ph. Eur. 1090601
Thymolblau Natriumsalz
Bestellinformation: Normally manufactured to order. Please check with customer services.
Formel:
C₂₇H₂₉NaO₅S Molecular Weight: 488,58 g/mol |
MDL:
MFCD00151093 CAS-Nummer: 62625-21-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...