Thymolphthalein 1 g/l in Ethanol Reag. Ph. Eur. 1090701

Lieferant: VWR Chemicals

Synonyme: 2-(1-Hydroxy-1,1-di(4-hydroxy-2-methyl-5-(methylethyl)phenyl)methyl)benzoesäurelacton, 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalid

87953.180EA 52.5 EUR
87953.180
Thymolphthalein 1 g/l in Ethanol Reag. Ph. Eur. 1090701
Thymolphthalein

Bestellinformation: Normally manufactured to order. Please check with customer services.

Formel: C₂₈H₃₀O₄
MDL: MFCD00005909
CAS-Nummer: 125-20-2
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR