4-Aminohippursäure Reag. Ph. Eur. 1003701

Lieferant: VWR Chemicals

Synonyme: p-Aminohippuric acid, 4-Aminohippuric Acid, 2-(4-Aminobenzamido)acetic acid, N-(4-Aminobenzoyl)glycine

87973.180EA 78.8 EUR
87973.180
4-Aminohippursäure Reag. Ph. Eur. 1003701
4-Aminohippursäure

Bestellinformation: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: H₂NC₆H₄C(O)NHCH₂COOH
Molecular Weight: 194,19 g/mol
MDL: MFCD00007890
CAS-Nummer: 61-78-9
EINECS: 000-000-0
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR