Tetraoctylammonium nitrate ≥99,0%, Selectophore™ Ionophor, Supelco®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
87983-500MG
87983-500MGEA
344
EUR
87983-500MG
Tetraoctylammonium nitrate ≥99,0%, Selectophore™ Ionophor, Supelco®
Tetraoctylammonium nitrate
Tetraoctylammonium nitrate (TOAN) is an ionophore or recognition element mostly used in potentiometric sensors.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
Formel:
[CH₃(CH₂)₆CH₂]₄N(NO₃) Molecular Weight: 528,89 g/mol |
MDL:
MFCD00525561 CAS-Nummer: 33734-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...