Tetraoctylammoniumbromid, Supelco®

Lieferant: Merck

Synonyme: Tetra-n-octylammonium bromide

M-Clarity™ Qualitätsstufe = MQ100

87994-5G
87994-5GEA 227 EUR
87994-5G
Tetraoctylammoniumbromid, Supelco®
Tetraoctylammoniumbromid
Formel: C₃₂H₆₈BrN
Molecular Weight: 546,8 g/mol
Schmelzpunkt: ~ 96 °C
Flammpunkt: 100 °C
MDL: MFCD00011863
CAS-Nummer: 14866-33-2
EINECS: 238-936-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83a692f542a87625f49dfc08e15dedceacd5532a&vid=85dbbcef79842a91e06e43cddad78d9c9a231cbe&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR