Tetraoctylammoniumbromid, Supelco®

Lieferant: Merck

Synonyme: Tetra-n-octylammonium bromide

M-Clarity™ Qualitätsstufe = MQ100

87996-10G-F
87996-10G-FEA 264 EUR
87996-10G-F
Tetraoctylammoniumbromid, Supelco®
Tetraoctylammoniumbromid
Formel: C₃₂H₆₈BrN
Molecular Weight: 546,8 g/mol
Schmelzpunkt: ~ 96 °C
Flammpunkt: 100 °C
MDL: MFCD00011863
CAS-Nummer: 14866-33-2
EINECS: 238-936-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c93275954c8b230fe74234327669abf5e2f0c85&vid=3a94129558d20108183cbd05c9b6ce5834fb29f3&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR