Tetrapentylammoniumbromid, Supelco®
Lieferant: Merck
|
Achtung
|
Synonyme:
Tetraamylammoniumbromid
M-Clarity™ Qualitätsstufe = MQ100
87997-10G-F
87997-10G-FEA
270
EUR
87997-10G-F
Tetrapentylammoniumbromid, Supelco®
Tetrapentylammoniumbromid
Formel:
C₂₀H₄₄BrN Molecular Weight: 378,48 g/mol Schmelzpunkt: 103 °C Dichte: 1,0662 g/cm³ (25 °C) |
MDL:
MFCD00011856 CAS-Nummer: 866-97-7 EINECS: 212-756-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...