Tetrapropylammoniumbromid, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

88103-10G
88103-10GEA 327 EUR
88103-10G
Tetrapropylammoniumbromid, Supelco®
Tetrapropylammoniumbromid
Formel: C₁₂H₂₈BrN
Schmelzpunkt: 265…274 °C
MDL: MFCD00011840
CAS-Nummer: 1941-30-6
EINECS: 217-727-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69517b799042487a6f090a997e50a823ee7e0073&vid=8e007de9a28a49f5ab3798f69efbb36482cc1312&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR