Methanol ≥99,5% (durch GC), GenAR® NF, Macron Fine Chemicals™

Lieferant: Avantor
Gefahr

Synonyme: Hydroxymethan, Methylalkohol, Holzalkohol

8818-06EA 0 EUR
8818-06 8818-14 8818-28
Methanol ≥99,5% (durch GC), GenAR® NF, Macron Fine Chemicals™
Methanol

GenAR—A line of reagents specifically developed for use in biotechnology and genetic research. 

Formel: CH₃OH
Siedepunkt: 64,6 °C (1013 hPa)
Schmelzpunkt: –98 °C
Dichte: 0,7918 g/cm³ (20 °C)
Flammpunkt: 12 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004595
CAS-Nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets E.P. Chemical Specifications
Meets N.F. Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay (CH₃0H) (by GC) ≥99.5 %
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Acidity Passes Test
NF - Alkalinity (as NH₃) ≤3 ppm
NF - Nonvolatile Residue ≤0.001 %
NF - Readily Carbonizable Substances Passes Test
NF - Readily Oxidizable Substances Passes Test
NF - Acetone, Aldehydes (as (CH₃)₂CO) ≤0.003 %
NF - Water (H₂O) ≤0.1 %
EP - Identification A Passes Test
EP - Identification B Passes Test
EP/BP - Appearance Passes Test
EP - Acidity or Alkalinity Passes Test
EP - Relative Density 0.791 - 0.793
EP - Absorbance at 230 nm (au) ≤0.15
EP - Absorbance at 250 nm (au) ≤0.050
EP - Absorbance at 270 nm (au) ≤0.020
EP - Absorbance at 290 nm (au) ≤0.010
EP - Scan from 230 nm to 290 nm Passes Test
EP/BP - Impurity A (Benzene) ≤2 ppm
EP - Related Substances - Single Impurity Peak ≤0.1 %
EP - Related Substances - Total of All Impurity Peaks ≤0.3 %
EP/BP - Reducing Substances Passes Test
EP/BP - Residue on Evaporation ≤10 ppm
EP - Water ≤0.10 %
Appearance (clear, colorless liquid)
Only Class 3 Solvents are likely to be present. Residual concentration
of all Class 3 Solvents is <0.5%.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR