Strontiumoxid ≥99,5% (Metall-Basis) SrO ≈97%, Pulver -100 Mesh

Lieferant: Thermo Scientific
Gefahr

88220.22
88220.22EA 581 EUR
88220.22
Strontiumoxid ≥99,5% (Metall-Basis) SrO ≈97%, Pulver -100 Mesh
Strontium oxide

Strontium oxide was used to make television picture tube glass. It finds application in glass, optic and ceramic industry. It is an active component of paint dryers. It is also used to prepare pure strontium up on heating with aluminum under vacuum.


Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water.

Formel: SrO
Molecular Weight: 103,62 g/mol
Siedepunkt: ∼3000 °C
Schmelzpunkt: 2430 °C
Dichte: 4,700
Lagertemperatur: Raumtemperatur
MDL: MFCD00049558
CAS-Nummer: 1314-11-0
EINECS: 215-219-9
UN: 3262
ADR: 8,II
Merck Index: 14,8846

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR