Quecksilber(II)tellurid ≥99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

88244.06
88244.06EA 248 EUR
88244.06
Quecksilber(II)tellurid ≥99% (Metall-Basis)
Quecksilber(II)tellurid

Purity: 99% CAS No.: 12068-90-5 Molecular Formula: HgTe Formula Weight: 328.19 Physical Form: -100 Mesh Powder Density: 8.63 Appearance: Gray to black Odour: Odourless Melting Point: 673° UN No.: 2025 MDL No.: MFCD00016133

Formel: HgTe
Schmelzpunkt: 673 °C
Dichte: 8,63
Lagertemperatur: Raumtemperatur
MDL: MFCD00016133
CAS-Nummer: 12068-90-5
EINECS: 235-108-9
UN: 2025
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR