Wasser, PESTINORM® für die Headspace-Gaschromatographie
Lieferant: VWR Chemicals
88251.290EA
57.8
EUR
88251.290
Wasser, PESTINORM® für die Headspace-Gaschromatographie
Wasser
Formel:
H₂O Siedepunkt: 100 °C (1013 hPa) Schmelzpunkt: 0 °C Dichte: 1 g/cm³ (4 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011332 CAS-Nummer: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Spezifikation Testergebnisse
Appearance | Clear colorless liquid |
Color (APHA, Pt-Co) | max. 5 |
Acidity (as Acetic acid) | max. 0.0005 % |
Alkalinity (as Ammonia) | max. 0.0001 % |
GC-HS (Each class 1 solvent) | max. 1 ug/g |
GC-HS (Each class 2 solvent) | max. 10 ug/g |
GC-HS (Each class 3 solvent) | max. 50 ug/g |
Headspace test for O.V.I's | Complying |
Suitable as solvent for the analysis of residual solvents of ICH classes 1, 2 and 3 according to Ph Eur and USP. |
|
Formic acid, acetic acid, N,N-dimethylacetamide, N,N-dimethylformamide and dimethyl sulfoxide are not detected |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...