Aluminum cobalt ≥99% (Metall-Basis), Pulver -100 mesh

Lieferant: Thermo Fisher Scientific
Gefahr

88320.36
88320.36EA 969 EUR
88320.36
Aluminum cobalt ≥99% (Metall-Basis), Pulver -100 mesh
Aluminum cobalt

Purity: 99% Molecular Formula: Al:Co; 69:31 wt% Odour: Odourless UN No.: UN3132 MDL No.: MFCD00239520

Formel: Al:Co;₆₉:₃₁wt%
Lagertemperatur: Raumtemperatur
MDL: MFCD00239520
UN: 3132
ADR: 4.3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR