Lithiumhydroxid, wasserfrei ≥99%, AnalaR NORMAPUR®
Lieferant: VWR Chemicals
|
Gefahr
|
88369.180EA
98.9
EUR
88369.180
88369.290
Lithiumhydroxid, wasserfrei ≥99%, AnalaR NORMAPUR®
Lithium hydroxide
Formel:
LiOH Molecular Weight: 23.95 g/mol Siedepunkt: 924 Schmelzpunkt: 462 |
MDL:
MFCD00011095 CAS-Nummer: 1310-65-2 UN: 2680 ADR: 8 II Merck Index: 13,05556 |
Spezifikation Testergebnisse
Assay | min. 99% |
Heavy metals (as Pb) | max. 20 ppm |
Lithium carbonate | max. 1.0% |
Cl (Chloride) | max. 0.04% |
SO4 (Sulphate) | max. 0.04% |
Ca (Calcium) | max. 0.02% |
Fe (Iron) | max. 20 ppm |
K (Potassium) | max. 0.02% |
Na (Sodium) | max. 0.05% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...