Salpetersäure 65% (max. 0.005 ppm Hg), AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

88371.320EA 170 EUR
88371.320 88371.290
Salpetersäure 65% (max. 0.005 ppm Hg), AnalaR NORMAPUR® analytisches Reagens
Salpetersäure
Formel: HNO₃
MDL: MFCD00011349
CAS-Nummer: 7697-37-2
UN: 2031
ADR: 8 (5.1),II

Bestellen

Spezifikation Testergebnisse

Assay min. 65%
Chloride (Cl) max. 0,5 ppm
Dithizone determination passes test
Mercury (Hg) max. 5 ppb
Residue after ignition max. 5 ppm
Sulphate (SO4) max. 1 ppm
Aluminium (Al) max. 0,05 ppm
Arsenic (As) max. 0,05 ppm
Barium (Ba) max. 0,02 ppm
Beryllium (Be) max. 0,01 ppm
Cadmium (Cd) max. 0,02 ppm
Calcium (Ca) max. 0,5 ppm
Chromium (Cr) max. 0,1 ppm
Cobalt (Co) max. 0,01 ppm
Copper (Cu) max. 0,01 ppm
Germanium (Ge) max. 0,05 ppm
Iron (Fe) max. 0,1 ppm
Lead (Pb) max. 0,05 ppm
Lithium (Li) max. 0,01 ppm
Magnesium (Mg) max. 0,1 ppm
Manganese (Mn) max. 0,01 ppm
Molybdenum (Mo) max. 0,02 ppm
Nickel (Ni) max. 0,05 ppm
Potassium (K) max. 0,1 ppm
Silver (Ag) max. 0,01 ppm
Sodium (Na) max. 0,5 ppm
Strontium (Sr) max. 0,01 ppm
Thallium (Tl) max. 0,05 ppm
Titanium (Ti) max. 0,1 ppm
Vanadium (V) max. 0,01 ppm
Zinc (Zn) max. 0,05 ppm
Zirconium (Zr) max. 0,1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR