Salzsäure 37% (max. 0.005 ppm Hg), AnalaR NORMAPUR® analytisches Reagens
Lieferant: VWR Chemicals
|
Gefahr
|
88372.320EA
78.1
EUR
88372.320
Salzsäure 37% (max. 0.005 ppm Hg), AnalaR NORMAPUR® analytisches Reagens
Salzsäure
Formel:
HCl |
MDL:
MFCD00011324 CAS-Nummer: 7647-01-0 UN: 1789 ADR: 8,II |
Spezifikation Testergebnisse
Assay | 37 - 38% |
Ammonium (NH4) | max. 3 ppm |
Free Chlorine | max. 0,5 ppm |
Mercury (Hg) | max. 5 ppb |
Residue after ignition | max. 5 ppm |
Sulphate (SO4) | max. 1 ppm |
Sulfite (SO3) | max. 2 ppm |
Aluminium (Al) | max. 0,05 ppm |
Arsenic (As) | max. 0,05 ppm |
Barium (Ba) | max. 0,02 ppm |
Beryllium (Be) | max. 0,01 ppm |
Cadmium (Cd) | max. 0,02 ppm |
Calcium (Ca) | max. 0,5 ppm |
Chromium (Cr) | max. 0,05 ppm |
Cobalt (Co) | max. 0,01 ppm |
Copper (Cu) | max. 0,01 ppm |
Germanium (Ge) | max. 0,1 ppm |
Iron (Fe) | max. 0,2 ppm |
Lead (Pb) | max. 0,05 ppm |
Lithium (Li) | max. 0,01 ppm |
Magnesium (Mg) | max. 0,05 ppm |
Manganese (Mn) | max. 0,01 ppm |
Molybdenum (Mo) | max. 0,02 ppm |
Nickel (Ni) | max. 0,05 ppm |
Potassium (K) | max. 0,1 ppm |
Silver (Ag) | max. 0,01 ppm |
Sodium (Na) | max. 0,3 ppm |
Strontium (Sr) | max. 0,01 ppm |
Thallium (Tl) | max. 0,05 ppm |
Titanium (Ti) | max. 0,05 ppm |
Vanadium (V) | max. 0,01 ppm |
Zinc (Zn) | max. 0,5 ppm |
Zirconium (Zr) | max. 0,05 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...