, HiPerSolv CHROMANORM® für die LC-MS
Lieferant: VWR Chemicals
88379.320EA
105
EUR
88379.320
, HiPerSolv CHROMANORM® für die LC-MS
Wasser mit 0,05% (v/v) Trifluoressigsäure
Spezifikation Testergebnisse
Nominal concentration (v/v) | 0.045 - 0.055% |
Evaporation residue | max. 0.0001% |
Ca (Calcium) | max. 0.2 ppm |
K (Potassium) | max. 0.2 ppm |
Mg (Magnesium) | max. 0.2 ppm |
Na (Sodium) | max. 0.5 ppm |
Transmittance (210 nm) | min. 20% |
Transmittance (220 nm) | min. 50% |
Transmittance (230 nm) | min. 80% |
Transmittance (240 nm) | min. 95% |
Transmittance (250 nm) | min. 99% |
Suitable for LC-MS | passes test |
Manufactured and filled under nitrogen |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...