Standards in Glasampullen für die Bestimmung des Kohlenwasserstoff-Index gemäß ISO 9377-2
Lieferant: VWR Chemicals
VWR® offers a complete range of products for the determination of the hydrocarbon oil index (HOI) in accordance with EN ISO 9377-2 (DIN H53).
- All standards are CRMs according to ISO 17025 and ISO 17034
- Shelf life of 24 months
- Storage: 2 – 8°C
- Supplied with a detailed, batch-specific certificate of analysis
The Hydrocarbon Oil Index (HOI) is defined as the total amount of compounds which can be extracted from the sample (drinking water, surface water and waste water) with a non-polar solvent having a boiling point between 39°C and 69°C .
In addition, the compounds must not absorb on Florisil® (magnesium sulphate) and must elute between n-decane (C10H24) and n-tetracontane (C40H82) when analyzed by GC using an apolar analytical column.
Standards: ISO 17025 / ISO 17034. ISO 9377-2.
Lieferumfang: Lieferung mit Analysenzertifikat, Bestell- und Chargennummer der Quellmaterialien.
Verpackung: In 1,0 und 10 ml-Glasampullen.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...