VWR®, Coulometry NewGen Reagent C-FA for coulometric Karl Fischer titration (10x5 ml)
Lieferant: VWR Chemicals
88424.005EA 390 EUR
88424.005
VWR®, Coulometry NewGen Reagent C-FA for coulometric Karl Fischer titration (10x5 ml)
Karl Fischer Titriermittel und Reagenzien
Catholyte for coulometric Karl Fischer titration of ketones and Li-ion battery electrolytes, Acetonitrile-based, free of alcohols (free of CMR substances).
  • free of alcohols
  • free of CMR substances
  • Recommended for Li-ion battery electrolytes
Specification:
Suitability (for KF-Titration): complying
Water content: 30 ppm