Ammoniumdihydrogenphosphat, Supelco®

Lieferant: Merck

Synonyme: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

M-Clarity™ Qualitätsstufe = MQ100

88431-25G
88431-25GEA 83.3 EUR
88431-25G
Ammoniumdihydrogenphosphat, Supelco®
Ammoniumdihydrogenphosphat
Formel: NH₄H₂PO₄
Molecular Weight: 115,03 g/mol
Schmelzpunkt: 190 °C
Dichte: 1,8 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003396
CAS-Nummer: 7722-76-1
EINECS: 231-764-5
Merck Index: 13,00547
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e861735254f68e2e865050d4618359c0ffe6b20b&vid=676f44d38c78718d056524383315e77ea2ec53a9&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR